Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Isosteviol, analytical standard, Supelco®
Organic reference standard, Isosteviol, analytical standard, Supelco®
Catalog # 92273-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Isosteviol, analytical standard, Supelco®
Catalog # 92273-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    27975-19-5
  • Description:
    Isosteviol, ≥98,0% (HPLC)
  • Formula:
    C₂₀H₃₀O₃
  • Description:
    Isosteviol
  • Form:
    Neat

Specifications

About this item