Organic reference standard, Isoeugenyl acetate, analytical standard, Supelco®
Katalog # 07055-100MG
Leverandør: Merck
CAS nummer:
Specifikationer
- Pakn:100 mg
- Standard:Analytical standard
- CAS-nr:93-29-8
- Beskrivelse:Isoeugenyl acetate
- Environmentally Preferable:
- Melting point:79...81 °C
- Formel:C₁₂H₁₄O₃
- Beskrivelse:Isoeugenyl acetate