Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Isobutyraldehyde, analytical standard, Supelco®
Organic reference standard, Isobutyraldehyde, analytical standard, Supelco®
Catalog # 68529-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Isobutyraldehyde, analytical standard, Supelco®
Catalog # 68529-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    78-84-2
  • Description:
    Isobutyraldehyde, ≥97,0% (GC)
  • Seq:
    01
  • Melting point:
    −65 °C (lit.)
  • Formula:
    C₄H₈O
  • Storage temperature:
    2...8 °C
  • Description:
    Isobutyraldehyde
  • Boiling point:
    63 °C (lit.)
  • Form:
    Neat

Specifications

About this item