Supelco® Organic Reference Standard, Ginkgolic acid C15:1
Catalog # 75741-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:22910-60-7
- Description:Ginkgolic acid C15:1
- Storage conditions:2…8 °C
- Hazard class description:H317 - H413
- Shipping conditions:STD
- MDL No.:MFCD03093717
- M-Clarity quality level:MQ100
- Synonym(s):22:1-ω7-Anacardic acid; 6-[(8Z)-Pentadecenyl]-salicylic acid; Anacardic acid monoene; Ginkgolic acid I; Romanicardic acid
- Molecular weight:346,50
- Flash point:265,5 °C - (calculated)
Specifications
About this item
- GHS signal: GHS07
Caution: P273 - P280 - P302 + P352