Organic reference standard, gamma-Valerolactone, analytical standard, Supelco®
40951-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:108-29-2
- Description:gamma-Valerolactone, ≥98,5% (GC)
- Melting point:−31 °C (lit.)
- Formula:C₅H₈O₂
- Description:gamma-Valerolactone
- Boiling point:207...208 °C (lit.); 82...85 °C/10 mmHg (lit.)
- Form:Neat