Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Fucoxanthin, analytical standard, Supelco®
Organic reference standard, Fucoxanthin, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Fucoxanthin, analytical standard, Supelco®
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    3351-86-8
  • Formula:
     C₄₂H₅₈O₆
  • Description:
    Fucoxanthin

Specifications

Related Information

Frequently Bought Together