Organic reference standard, Farnesol, analytical standard, Supelco®
43348-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:4602-84-0
- Description:Farnesol, ≥95,0% (GC)
- Formula:C₁₅H₂₆O
- Storage temperature:2...8 °C
- Description:Farnesol
- Boiling point:149 °C/4 mmHg (lit.)
- Form:Neat