Organic reference standard, ethyl hexanoate, analytical standard, Supelco®
Catalog # 08375-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:123-66-0
- Description:Ethyl hexanoate, ≥99,5% (GC)
- Formula:C₈H₁₆O₂
- Description:Ethyl hexanoate
- Boiling point:168 °C (lit.)