Supelco® Organic Reference Standard, Ellagic Acid
14668-50MG
: Merck
:
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:476-66-4
- Description:Ellagic acid
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00006914
- M-Clarity quality level:MQ100
- Synonym(s):4,4′,5,5′,6,6′-Hexahydroxydiphenic acid 2,6,2′,6′-dilactone; Alizarin Yellow; Benzoaric acid; Elagostasine; Eleagic acid; Gallogen; Lagistase
- Formula:C₁₄H₆O₈
- Molecular weight:302,19
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.
: P264 - P270 - P301 + P312 - P501