Supelco® Organic Reference Standard, Eleutheroside E
Eleutheroside E, Analytical standard, 10 mg
Catalog # 08198-10MG
Supplier: Merck
Supelco® Organic Reference Standard, Eleutheroside E
Catalog # 08198-10MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
- GHS signal: GHS07
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Specifications
Specifications
- Pk:10 mg
- Standards:Analytical standard
- Description:Eleutheroside E
- Cat. No.:08198-10MG
- M-Clarity quality level:MQ100
- Formula:C₃₄H₄₆O₁₈
- Storage conditions:2...8 °C
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Synonym(s):[(1R,3aR,4S,6aS)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2,6-dimethoxy-4,1-phenylene) bis-β-ᴅ-glucopyranoside
- Molecular weight:742,72
- CAS No.:39432-56-9
- Shipping conditions:STD



