Supelco® Organic Reference Standard, DL-Homocysteine
44925-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:454-29-5
- Description:ᴅʟ-Homocysteine
- Storage conditions:–20 °C
- Shipping conditions:STD
- MDL No.:MFCD00004898
- M-Clarity quality level:MQ100
- Synonym(s):2-Amino-4-mercaptobutyric acid
- Melting point:232...233 °C (lit.)
- Formula:HSCH₂CH₂CH(NH₂)COOH
- Molecular weight:135,18
- Storage temperature:−20 °C
- Flash point:Not applicable
- Form:Neat
: Store under argon.