Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Dioctyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Organic reference standard, Dioctyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Catalog # 34193-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Dioctyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Catalog # 34193-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    93952-13-7
  • Description:
    Dioctyl phthalate-3,4,5,6-d4
  • Melting point:
    –50 °C (lit.)
  • Formula:
    C₂₄H₃₈O₄
  • Description:
    Dioctyl phthalate-3,4,5,6-d4
  • Boiling point:
    384 °C (lit.)
  • Form:
    Neat

Specifications

About this item