Organic reference standard, Dimethyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
34189-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:93951-89-4
- Description:Dimethyl phthalate-3,4,5,6-d4
- Melting point:2 °C (lit.)
- Formula:C₁₀H₁₀O₄
- Description:Dimethyl phthalate-3,4,5,6-d4
- Boiling point:282 °C (lit.)
- Form:Neat