Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Dimethyl disulfide, analytical standard, Supelco®
Organic reference standard, Dimethyl disulfide, analytical standard, Supelco®
Catalog # 68986-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Dimethyl disulfide, analytical standard, Supelco®
Catalog # 68986-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    624-92-0
  • Description:
    Dimethyl disulfide, ≥98,5% (GC)
  • Melting point:
    –85 °C (lit.)
  • Formula:
    C₂H₆S₂
  • Description:
    Dimethyl disulfide
  • Boiling point:
    109 °C (lit.)
  • Form:
    Neat

Specifications

About this item