Organic reference standard, Dihydrocoumarin, analytical standard, Supelco®
07552-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:119-84-6
- Description:Dihydrocoumarin, ≥98,5% (GC)
- Melting point:24...25 °C (lit.)
- Formula:C₉H₈O₂
- Description:Dihydrocoumarin
- Boiling point:272 °C (lit.)