Organic reference standard, delta-Damascone, analytical standard, Supelco®
Catalog # 55137-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:57378-68-4
- Description:delta-Damascone, ≥90,0% (GC)
- Environmentally Preferable:
- Formula:C₁₃H₂₀O
- Description:delta-Damascone