Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, D-(+)-Sucrose octaacetate, analytical standard, Supelco®
Organic reference standard, D-(+)-Sucrose octaacetate, analytical standard, Supelco®
  94474-100MG
 :  Merck
 :  
undefined
Organic reference standard, D-(+)-Sucrose octaacetate, analytical standard, Supelco®
  94474-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    126-14-7
  • Description:
    D-(+)-Sucrose octaacetate, ≥98,0% (TLC)
  • Melting point:
    82...85 °C (lit.)
  • Formula:
    C₂₈H₃₈O₁₉
  • Description:
    D-(+)-Sucrose octaacetate
  • Boiling point:
    260 °C (lit.)
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.