Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, coenzyme Q10, analytical standard, Supelco®
Organic reference standard, coenzyme Q10, analytical standard, Supelco®
Catalog # 07386-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, coenzyme Q10, analytical standard, Supelco®
Catalog # 07386-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    303-98-0
  • Description:
    Coenzyme Q10, ≥97.5%
  • Melting point:
    49 °C
  • Formula:
    C₅₉H₉₀O₄
  • Storage temperature:
    −20 °C
  • Description:
    Coenzyme Q10

Specifications

About this item

Caution: Light sensitive