Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, cis-3-Hexen-1-ol, analytical standard, Supelco®
Organic reference standard, cis-3-Hexen-1-ol, analytical standard, Supelco®
Catalog # 91316-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, cis-3-Hexen-1-ol, analytical standard, Supelco®
Catalog # 91316-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    928-96-1
  • Description:
    cis-3-Hexen-1-ol, ≥98,0% (GC)
  • Formula:
    C₆H₁₂O  
  • Description:
    cis-3-Hexen-1-ol
  • Boiling point:
    156...157 °C(lit.)
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.