Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, chrysoidine G, analytical standard, Supelco®
Organic reference standard, chrysoidine G, analytical standard, Supelco®
Catalog # 80028-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, chrysoidine G, analytical standard, Supelco®
Catalog # 80028-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    532-82-1
  • Description:
    Chrysoidine , ≥95,0% (HPLC)
  • Melting point:
    235 °C (dec.) (lit.)
  • Formula:
    C₁₂H₁₂N₄ ·HCl
  • Storage temperature:
    2...8 °C
  • Description:
    Chrysoidine G
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.