Supelco® Organic Reference Standard, Chrysin
95082-50MG
: Merck
:
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:480-40-0
- Description:Chrysin
- Environmentally Preferable:
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00006834
- M-Clarity quality level:MQ100
- Synonym(s):5,7-Dihydroxyflavone
- Melting point:284 - 286 °C (lit.)
- Molecular weight:254,24