Supelco® Organic Reference Standard, Carvacrol
42632-50MG
: Merck
:
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:499-75-2
- Description:Carvacrol
- Storage conditions:2...8 °C
- Hazard class description:H302 - H314 - H411
- Shipping conditions:STD
- MDL No.:MFCD00002236
- M-Clarity quality level:MQ100
- Synonym(s):5-Isopropyl-2-methylphenol
- Melting point:3...4 °C (lit.)
- Formula:(CH₃)₂CHC₆H₃(CH₃)OH
- Molecular weight:150,22
- Storage temperature:2...8 °C
- Density:0,976 g/ml at 20 °C (lit.)
- Flash point:106 °C - closed cup
- Boiling point:236...237 °C (lit.)
- Form:Neat
- GHS signal: GHS05, GHS07
: Bottomless glass bottle. Contents are inside inserted fused cone.
: P273 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338