Organic reference standard, Butylamine, analytical standard, Supelco®
Catalog # 90893-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:109-73-9
- Description:Butylamine, ≥99,0% (GC)
- Seq:01
- Melting point:–49 °C (lit.)
- Formula:C₄H₁₁N
- Description:Butylamine
- Boiling point:78 °C (lit.)
- Form:Neat