Organic reference standard, Bergapten, analytical standard, Supelco®
Catalog # 69664-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:484-20-8
- Description:Bergapten, ≥99,0% (HPLC)
- Melting point:189...193 °C; 190...193 °C (lit.)
- Formula:C₁₂H₈O₄
- Storage temperature:2...8 °C
- Description:Bergapten
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle contents are inside inserted with fused cone.