Supelco® Organic Reference Standard, Bacoside A3
Catalog # 53889-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:157408-08-7
- Description:Bacoside A3
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD07779137
- M-Clarity quality level:MQ100
- Synonym(s):(3β,16β,23R)-16,23:16,30-Diepoxy-20-hydroxydammar-24-en-3-yl O-α-ʟ-arabinofuranosyl-(1→2)-O-[β-ᴅ-glucopyranosyl-(1→3)]-β-ᴅ-glucopyranoside; Jujubogenin 3-O-[α-ʟ-arabinofuranosyl-(1→2)-O-[β-ᴅ-glucopyranosyl-(1→3)]-β-ᴅ-glucopyranoside]
- Molecular weight:929,10