Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, Apigenin 7-Glucoside
Supelco® Organic Reference Standard, Apigenin 7-Glucoside
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Apigenin 7-Glucoside
Supplier:  Merck

Specifications

  • CAS No.:
    578-74-5
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00016787
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4′,5,7-Trihydroxyflavone 7-glucoside; Apigetrin; Cosmosiin
  • Formula:
    C₂₁H₂₀O₁₀
  • Molecular weight:
    432,38
  • Flash point:
    Not applicable

Specifications

Frequently Bought Together