Order Entry
Denmark
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Andrographolide
  90281-5MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Andrographolide
  90281-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    5508-58-7
  • Description:
    Andrographolide
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD07778082
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3E,4S)-3-[2-[(1R,4aS,5R,6R,8aS)-Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethylidene]dihydro-4-hydroxy-2(3H)-furanone; ent-(3β,12E,14R)-3,14,19-Trihydroxy-8(17),12-labdadien-16,15-olide; Andrographis
  • Molecular weight:
    350,45