Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, Amino Acid Standards, Physiological
Supelco® Organic Reference Standard, Amino Acid Standards, Physiological
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Amino Acid Standards, Physiological
Supplier:  Merck
Certificates

Specifications

  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H290
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00162052

Specifications

Related Information

Frequently Bought Together