Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, alpha-Hexylcinnamaldehyde, analytical standard, Supelco®
Organic reference standard, alpha-Hexylcinnamaldehyde, analytical standard, Supelco®
Catalog # 09178-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, alpha-Hexylcinnamaldehyde, analytical standard, Supelco®
Catalog # 09178-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    101-86-0
  • Description:
    alpha-Hexylcinnamaldehyde, ≥98,5% (GC)
  • Formula:
    C₁₅H₂₀O
  • Description:
    alpha-Hexylcinnamaldehyde
  • Boiling point:
    174...176 °C/15 mmHg (lit.)

Specifications

About this item