Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, 7-Methylcoumarin, analytical standard, Supelco®
  14673-100MG
 :  Merck
 :  
undefined
Organic reference standard, 7-Methylcoumarin, analytical standard, Supelco®
  14673-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2445-83-2
  • Description:
    7-Methylcoumarin, ≥98,0% (HPLC)
  • Melting point:
    128...130 °C (lit.)
  • Formula:
    C₁₀H₈O₂
  • Description:
    7-Methylcoumarin
  • Boiling point:
    171...172 °C/11 mm Hg (lit.)