Organic reference standard, 5-Tricosylresorcinol, analytical standard, Supelco®
Catalog # 03422-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:70110-60-0
- Description:5-Tricosylresorcinol, ≥95,0% (HPLC)
- Formula:C₂₉H₅₂O₂
- Storage temperature:2...8 °C
- Description:5-Tricosylresorcinol
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.