Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 5-Methylfurfural, analytical standard, Supelco®
Organic reference standard, 5-Methylfurfural, analytical standard, Supelco®
Catalog # 91307-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 5-Methylfurfural, analytical standard, Supelco®
Catalog # 91307-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    620-02-0
  • Description:
    5-Methylfurfural, ≥98,5% (GC)
  • Formula:
    C₆H₆O₂
  • Description:
    5-Methylfurfural
  • Flash point:
    72 °C
  • Boiling point:
    187 °C (lit.)
  • Form:
    Neat

Specifications

About this item