Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 5-Heneicosylresorcinol, analytical standard, Supelco®
Organic reference standard, 5-Heneicosylresorcinol, analytical standard, Supelco®
  50851-10MG
 :  Merck
 :  
undefined
Organic reference standard, 5-Heneicosylresorcinol, analytical standard, Supelco®
  50851-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    70110-59-7
  • Description:
    5-Heneicosylresorcinol, ≥95,0% (HPLC)
  • Formula:
    C₂₇H₄₈O₂
  • Storage temperature:
    2…8 °C
  • Description:
    5-Heneicosylresorcinol
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.