Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, 4-Methyl-5-thiazoleethanol, analytical standard, Supelco®
  61352-100MG
 :  Merck
 :  
undefined
Organic reference standard, 4-Methyl-5-thiazoleethanol, analytical standard, Supelco®
  61352-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    137-00-8
  • Description:
    4-Methyl-5-thiazoleethanol, ≥98,0% (GC)
  • Formula:
    C₆H₉NOS
  • Description:
    4-Methyl-5-thiazoleethanol
  • Flash point:
    112 °C
  • Boiling point:
    135 °C/7 mm Hg (lit.)
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.