Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 4-Methoxyphenol, analytical standard, Supelco®
Organic reference standard, 4-Methoxyphenol, analytical standard, Supelco®
Catalog # 92264-250MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 4-Methoxyphenol, analytical standard, Supelco®
Catalog # 92264-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    150-76-5
  • Description:
    4-Methoxyphenol, ≥98,5% (GC)
  • Melting point:
    55...57 °C (lit.)
  • Formula:
    C₇H₈O₂
  • Description:
    4-Methoxyphenol
  • Flash point:
    132 °C
  • Boiling point:
    243 °C (lit.)
  • Form:
    Neat

Specifications

About this item