Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, 4-Hydroxybenzaldehyde, analytical standard, Supelco®
  91554-250MG
 :  Merck
 :  
undefined
Organic reference standard, 4-Hydroxybenzaldehyde, analytical standard, Supelco®
  91554-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    123-08-0
  • Description:
    4-Hydroxybenzaldehyde, ≥97,5% (GC)
  • Melting point:
    112...116 °C (lit.)
  • Formula:
    C₇H₆O₂
  • Description:
    4-Hydroxybenzaldehyde
  • Form:
    Neat