Organic reference standard, 4'-Hydroxyacetophenone, analytical standard, Supelco®
Catalog # 98837-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:99-93-4
- Description:4'-Hydroxyacetophenone, ≥98,0% (GC)
- Melting point:107…110 °C; 109...111 °C (lit.)
- Formula:C₈H₈O₂
- Description:4'-Hydroxyacetophenone
- Boiling point:147...148 °C/3 mm Hg (lit.)
- Form:Neat