Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 3-Carene, analytical standard, Supelco®
Organic reference standard, 3-Carene, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, 3-Carene, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    13466-78-9
  • Formula:
    C₁₀H₁₆
  • Description:
    3-Carene
  • Flash point:
    47 °C
  • Boiling point:
    168...169 °C/705 mm Hg (lit.)

Specifications

Frequently Bought Together