Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, 3β-Taraxerol
Supelco® Organic Reference Standard, 3β-Taraxerol
Catalog # 75934-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 3β-Taraxerol
Catalog # 75934-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    127-22-0
  • Description:
    3β-Taraxerol
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD13186859
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3β)-ᴅ-Friedoolean-14-en-3-ol; (3β13α)-13-Methyl-27-norolean-14-en-3-ol; Alnulin; Skimmiol; Taraxerol; Tiliadin
  • Molecular weight:
    426,72

Specifications

About this item