Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 2-Undecanone, analytical standard, Supelco®
Organic reference standard, 2-Undecanone, analytical standard, Supelco®
Catalog # 74038-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2-Undecanone, analytical standard, Supelco®
Catalog # 74038-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    112-12-9
  • Description:
    2-Undecanone, ≥98,0% (GC)
  • Melting point:
    11...13 °C (lit.)
  • Formula:
    C₁₁H₂₂O
  • Description:
    2-Undecanone
  • Boiling point:
    231...232 °C (lit.)

Specifications

About this item