Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 2-Pentanol, analytical standard, Supelco®
Organic reference standard, 2-Pentanol, analytical standard, Supelco®
Catalog # 44759-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2-Pentanol, analytical standard, Supelco®
Catalog # 44759-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    6032-29-7
  • Description:
    2-Pentanol, ≥97,0% (GC)
  • Formula:
    C₅H₁₂O
  • Description:
    2-Pentanol
  • Boiling point:
    118...119 °C (lit.)

Specifications

About this item