Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 2-Methylfuran, analytical standard, Supelco®
Organic reference standard, 2-Methylfuran, analytical standard, Supelco®
Catalog # 39587-5ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2-Methylfuran, analytical standard, Supelco®
Catalog # 39587-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    534-22-5
  • Description:
    2-Methylfuran, ≥99,0% (GC)
  • Seq:
    02
  • Formula:
    C₅H₆O
  • Description:
    2-Methylfuran
  • Flash point:
    –22 °C
  • Boiling point:
    63...66 °C (lit.)
  • Form:
    Neat

Specifications

About this item