Organic reference standard, 2-Furanmethanethiol, analytical standard, Supelco®
Catalog # 67319-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:98-02-2
- Description:2-Furanmethanethiol, ≥98,0% (HPLC)
- Formula:C₅H₆OS
- Storage temperature:2…8 °C
- Description:2-Furanmethanethiol
- Flash point:45 °C
- Boiling point:155 °C
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.