Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 1-Dodecanol, analytical standard, Supelco®
Organic reference standard, 1-Dodecanol, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, 1-Dodecanol, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    112-53-8
  • Melting point:
    22...26 °C (lit.)
  • Formula:
    C₁₂H₂₆O
  • Description:
    1-Dodecanol
  • Flash point:
    121 °C
  • Boiling point:
    260...262 °C (lit.)

Specifications

Frequently Bought Together