Supelco® Organic Reference Standard, [6]-Gingerol
50866-10MG
: Merck
:
- Pakn:10 mg
- Standard:Analytical standard
- CAS-nr:23513-14-6
- Beskrivelse:[6]-Gingerol
- Environmentally Preferable:
- Storage conditions:2…8 °C
- Fareklassebeskrivelse:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00210507
- M-Clarity quality level:MQ100
- Synonym(s):3-Decanone, 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (5S)-; 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone; 6-Gingerol
- Molecular weight:294,39
- GHS signal: GHS06
: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338