Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, (+/-)-2-Octanol, analytical standard, Supelco®
Organic reference standard, (+/-)-2-Octanol, analytical standard, Supelco®
Catalog # 42277-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, (+/-)-2-Octanol, analytical standard, Supelco®
Catalog # 42277-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    123-96-6
  • Description:
    (+/-)-2-Octanol, ≥97,0% (GC)
  • Formula:
    C₁₀H₁₉NO₃
  • Description:
    (+/-)-2-Octanol
  • Form:
    Neat

Specifications

About this item