Supelco® Organic Reference Standard, (−)-Bilobalide
Catalog # 79593-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:33570-04-6
- Description:(−)-Bilobalide
- Storage conditions:2…8 °C
- Shipping conditions:WICE
- MDL No.:MFCD00132880
- M-Clarity quality level:MQ100
- Synonym(s):Bilobalide
- Molecular weight:326,30