Order Entry
Denmark
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-β-Elemene

Supelco® Organic Reference Standard, (−)-β-Elemene

(−)-β-Elemene, Analytical standard, 25 mg
Catalog # 63965-25MG
Supplier:   Merck
Supelco® Organic Reference Standard, (−)-β-Elemene
Catalog # 63965-25MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, (−)-β-Elemene
Supelco® Organic Reference Standard, (−)-β-Elemene

(−)-β-Elemene, Analytical standard, 25 mg

CAS Number:  
Price
3.260,00 kr
Log in for availability
1 * 25 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • Description:
    (−)-β-Elemene
  • Cat. No.:
    63965-25MG
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2…8 °C
  • MDL No.:
    MFCD00468041
  • Flash point:
    98 °C
  • Synonym(s):
    β-Elemene; (1S,2S,4R)-(−)-2,4-Diisopropenyl-1-methyl-1-vinylcyclohexane; (1S,2S,4R)-1-Ethenyl-1-methyl-2,4-bis(1-methylethenyl)cyclohexane
  • Molecular weight:
    204,35
  • CAS No.:
    515-13-9
  • Shipping conditions:
    STD

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.