Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Chicoric Acid
Supelco® Organic Reference Standard, (−)-Chicoric Acid
Catalog # 06957-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (−)-Chicoric Acid
Catalog # 06957-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    70831-56-0
  • Description:
    (−)-Chicoric acid
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD22683653
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2R,3R)-2,3-Bis{[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy}butanedioic acid; 2,3-Di-trans-caffeoyltartaric acid
  • Formula:
    C₂₂H₁₈O₁₂
  • Molecular weight:
    474,37
  • Storage temperature:
    2…8 °C
  • Flash point:
    Not applicable

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.