JC-1 iodide salt mitochondria dye, suitable for fluorescence
Catalog # 70014.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:652.23 g/mol
Storage Temperature:Refrigerator
CAS Number:47729-63-5
:3520-43-2
Formula:C₂₅H₂₇Cl₄IN₄
MDL Number:MFCD00188475